VWR®, Glasperlen für das Mahlen und Homogenisieren
Lieferant: VWR Collection
432-0064EA 324 EUR
432-0064
VWR®, Glasperlen für das Mahlen und Homogenisieren
Kugeln Glaskugeln
Small beads suitable for use with vortex mixers and tube homogenisers.
Molecular biology grade beads are treated to be free if DNase and RNase, making them suitable for a range of applications including disruption of mold and yeasts.