Grease, high vacuum, Dow Corning®
Lieferant: Thermo Scientific
44224.KT
44224.KTEA 82.2 EUR
44224.KT
Grease, high vacuum, Dow Corning®
Schmiermittel
High-vacuum grease is used as an anti-seize grease in ground glass joints and to lubricate synthetic and silicone rubber gaskets. Also used to lubricate water-treating equipment, laboratory equipment, lubricate O-rings on binoculars and telescopes and to prevent fogging of delicate lenses.
  • Recommended for vacuums reaching between 1×10-5 and 1×10-6 mmHg
  • Immiscible with water, methanol, ethanol, acetone, acetone, glycol and glycerine
  • Incompatible with oxidising agents