L-Methionin, Millipore®

Lieferant: Merck Millipore Calbiochem

Synonyme: H-Met-OH, (S)-(-)-2-Amino-4-(methylmercapto)buttersäure, (S)-(-)-2-Amino-4-(methylthio)buttersäure, L(-)-Methionin, (S)-(-)-Methionin

4500-100
4500-100EA 91.8 EUR
4500-100
L-Methionin, Millipore®
L-Methionin
Formel: C₅H₁₁NO₂S
Molecular Weight: 149,21 g/mol
Siedepunkt: 278 °C (1013 hPa)
Schmelzpunkt: 136 °C
Dichte: 1,373 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00063097
CAS-Nummer: 63-68-3
EINECS: 200-562-9
Merck Index: 13,06004

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR