Program controller
Lieferant: GESTIGKEIT HARRY
PR5-3T
453-2109EA 2347 EUR
453-2109
Program controller
Regler Temperaturregler
The micro-processor controlled ramp controller is a complex time/temperature control and this program is indefinitely stored by means of an EEPROM.
  • Pogramming enabling each value to be viewed for control purposes
  • Programmable pre-selected constant time makes it active at any time
  • Each temperature and time parameter can be selected with their own keys
Suitable for the high-temperature hotplate.