Flask, Digesdahl®
Lieferant: Hach
2312542
458-0048EA 139 EUR
458-0048
Flask, Digesdahl®
Kolben Stehkolben
Specially designed glass flask for use with Digesdahl® digestion apparatus. Each flask is calibrated individually with a 100 ml (to contain) mark.