Ion exchange resins, Diaion® PA312 (Cl)
Lieferant: Thermo Scientific
46426.A1
46426.A1EA
81.5
EUR
46426.A1
Ion exchange resins, Diaion® PA312 (Cl)
Chromatographie-Sorbentien
HPLC-Sorbentien
These resins are ideal for use in decolourisation in sugar solutions, metal recovery, pharmaceuticals and purification of amino acids.