Yttrium, oil based standard solution, Y 1000 µg/g, Specpure®
Lieferant: Thermo Scientific
47172.18
47172.18EA
114
EUR
47172.18
Yttrium, oil based standard solution, Y 1000 µg/g, Specpure®
Standards
Kationischer Standards
Yttrium-Standards
Physical Form: Liquid
MDL No.: MFCD00011468
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...