Miracloth, Calbiochem®
Lieferant: Merck Millipore (Calbiochem‎)
475855-1
475855-1EA 189 EUR
475855-1
Miracloth, Calbiochem®
Filter Membranfilter
Miracloth is an autoclavable, quick filtration material for gelatinous homogenates that is composed of rayon-polyester with an acrylic binder. Ideal for filtration of cleared neutralised lysates during medium- and large-scale plasmid purification to prevent contamination with loosely adherent pellet material and is useful for protoplast isolation.