NAD+ lithium salt ≥90% (durch enzymatische Reinheit), Calbiochem®

Lieferant: Merck Millipore Calbiochem

Synonyme: β-Nicotinamide-adenine Dinucleotide, Li

481915-1
481915-1EA 140 EUR
481915-1
NAD+ lithium salt ≥90% (durch enzymatische Reinheit), Calbiochem®
NAD+ lithium salt

A major electron acceptor molecule in biological oxidations.

Formel: C₂₁H₂₆N₇O₁₄P₂Li
Molecular Weight: 669,4 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 64417-72-7
Merck Index: 14,06344

Bestellen

Spezifikation Testergebnisse

Purity (by Enzymatic Assay) ≥90%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR