Kaliumhydrogenphthalat 99,95-100,05% (Trockenmasse), ULTREX®, ultrarein, J.T.Baker®
Lieferant: Avantor
Synonyme:
Phthalsäure Kaliumsalz, 1,2-Benzoldicarbonsäure Kaliumsalz, Kaliumphthalat monobasisch
4889-03EA
706.49
EUR
4889-03
Kaliumhydrogenphthalat 99,95-100,05% (Trockenmasse), ULTREX®, ultrarein, J.T.Baker®
Kaliumhydrogenphthalat
For primary standard and research applications.
Formel:
HOOCC₆H₄COOK Molecular Weight: 204,22 g/mol Schmelzpunkt: 295…300 °C Dichte: 1,64 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013070 CAS-Nummer: 877-24-7 EINECS: 212-889-4 Merck Index: 13,07694 |
Spezifikation Testergebnisse
For Laboratory,Research,or Manufacturing Use | 99.95 - 100.05% |
Assay (1-KOCOC₆H₄-2-COOH) (dried basis) | 99.95 - 100.05 % |
Identification (by IR) | Passes Test |
Particulate Matter, For Information Only | |
pH of 0.05M Solution at 25°C | 4.00 - 4.02 |
Non-Metallic Impurities - Chlorine Compounds (as Cl), For Information Only | |
Non-Metallic Impurities - Silicon (Si), For Information Only | |
Non-Metallic Impurities - Sulfur Compounds (as S), For Information Only | |
Metallic Impurities - Aluminum (Al), For Information Only | |
Metallic Impurities - Barium (Ba), For Information Only | |
Metallic Impurities - Bismuth (Bi), For Information Only | |
Metallic Impurities - Cadmium (Cd), For Information Only | |
Metallic Impurities - Calcium (Ca), For Information Only | |
Metallic Impurities - Chromium (Cr), For Information Only | |
Metallic Impurities - Cobalt (Co), For Information Only | |
Metallic Impurities - Copper (Cu), For Information Only | |
Metallic Impurities - Iron (Fe), For Information Only | |
Metallic Impurities - Lead (Pb), For Information Only | |
Metallic Impurities - Magnesium (Mg), For Information Only | |
Metallic Impurities - Manganese (Mn), For Information Only | |
Metallic Impurities - Mercury (Hg), For Information Only | |
Metallic Impurities - Molybdenum (Mo), For Information Only | |
Metallic Impurities - Nickel (Ni), For Information Only | |
Metallic Impurities - Niobium (Nb), For Information Only | |
Metallic Impurities - Silver (Ag), For Information Only | |
Metallic Impurities - Sodium (Na), For Information Only | |
Metallic Impurities - Strontium (Sr), For Information Only | |
Metallic Impurities - Tin (Sn), For Information Only | |
Metallic Impurities - Titanium (Ti), For Information Only | |
Metallic Impurities - Vanadium (V), For Information Only | |
Metallic Impurities - Zinc (Zn), For Information Only | |
Metallic Impurities - Zirconium (Zr), For Information Only | |
Certificate Provided Reports Actual Lot Analysis |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...