2,6-Pyridindicarbonsäure 0.02 M in Wasser Eluent-Konzentrat, für die Ionenchromatographie

Lieferant: Merck

Synonyme: Dipicolinsäure, DPA

M-Clarity™ Qualitätsstufe = MQ100

50972-1L
50972-1LEA 58.7 EUR
50972-1L
2,6-Pyridindicarbonsäure 0.02 M in Wasser Eluent-Konzentrat, für die Ionenchromatographie
2,6-Pyridindicarbonsäure

This certified eluent concentrate for ion chromatography is traceable by potentiometric titration to NIST Standard Reference Material.


Prepared with 2,6-pyridinecarboxylic acid and high purity water (18.2 MΩ, 0.2 μm filtered)


Standards: It is certified in accordance with ISO Guide 31.

Formel: C₇H₅NO₄
Molecular Weight: 167,12 g/mol
MDL: MFCD00006299
CAS-Nummer: 499-83-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4944799031b84af2c8194119dfb246938aec65&vid=23f9715bf7de3eb8269f00f983e49e60e60242dc&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR