Blood stain card, Whatman™
Lieferant: QIAGEN
WB100014
512-1150EA 0 EUR
512-1150
Blood stain card, Whatman™
Nukleinsäure-Aufbewahrungssysteme
For the collection and transport of blood and body fluids.
  • Made from absorbent filter paper
  • Enables efficient sample collection
  • Features a protective cover that aids sample drying and protects sample from direct contact
  • Designed for short-term handling of specimens