Forceps for membrane filters
Lieferant: Sartorius
16625
512-3304EA 40.2 EUR
512-3304
Forceps for membrane filters
Pinzetten
These filter forceps are made of high grade stainless steel.
  • Autoclavable, can be flamed
  • With smooth, flat tips for a careful, firm hold of the membrane filter
Membrane filters should only be handled with suitable tweezers in order to avoid contamination which can result from hand contact.