Phosphorus standard for AAS, TracecERT®, Supelco®
Lieferant: Merck
51474-250ML
51474-250MLEA
37.5
EUR
51474-250ML
Phosphorus standard for AAS, TracecERT®, Supelco®
Standards
Kationischer Standards
Phosphor-Standards
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034