Culture Capsules, 0.2 µm, Sterile
Lieferant: Cytiva (Formerly Pall Lab)
4227
516-7587EA 137 EUR
516-7587
Culture Capsules, 0.2 µm, Sterile
Filter Membranfilterkapseln
Specifically designed for sterilizing cell culture media.
  • Disposable
  • Two layers, Non-pyrogenic
  • Pleated design for maximum throughput and effective filtration
Disposable culture capsules incorporate two layers of low protein binding HT Tuffryn® (polysulfone) membrane for reliable purification.
Body housing and end caps are constructed of polycarbonate with a polypropylene core and vent plug. The upstream/downstream filter support is polyester. For additional safety, a polycarbonate filling bell is available. Sterilized by gamma irradiation. If further sterilization is desired prior to use, capsules may be autoclaved once only at 121 °C (250 °F) for 20 minutes at 1,0 bar (15 psi).
Culture capsules pass USP Class VI-121°C Plastics Test. Endotoxicity is less than 0,5 EU/mL using the Limulus Amebocyte Lysate (LAL) Test. Finished product has been sampled and tested for bacterial retention per ASTM F838-83 methodology using Brevundimonous diminuta at 107 organisms/cm² in cell culture media.
Minimum Bubble Point – Water: 2,4 bar (240 kPa, 35 psi)
Bestellinformation: Supplied individually wrapped.