Leichtdestillat ≥99,9995% (Metall-Basis) ICP solvent für anorganische Spurenanalyse, Supelco®
Lieferant: Merck
|
Gefahr
|
54362-1L
54362-1LEA
0
EUR
54362-1L
Leichtdestillat ≥99,9995% (Metall-Basis) ICP solvent für anorganische Spurenanalyse, Supelco®
Leichtdestillat
ICP solvent is low odor and reduced toxicity equivalent to classical kerosene or xylene and ideal for dilution of oils and other organic fluids for determination of metal traces.
Siedepunkt:
200…250 °C (Lit.) |
CAS-Nummer:
64742-47-8 UN: 1993 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...