Pyridoxal Hydrochlorid, Millipore®

Lieferant: Merck Millipore Calbiochem

Synonyme: 3-Hydroxy-5-(hydroxymethyl)-2-methylisonicotinaldehyde hydrochloride, Pyridoxal HCl

545060-25
545060-25EA 299 EUR
545060-25
Pyridoxal Hydrochlorid, Millipore®
Pyridoxal Hydrochlorid
Formel: C₈H₁₀ClNO₃
Molecular Weight: 203,62 g/mol
MDL: MFCD00012809
CAS-Nummer: 65-22-5
EINECS: 200-602-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR