TLC education advanced kit, micro set F1
Lieferant: MACHEREY-NAGEL
814200 814201 814202 814203 814204 814205 814206
552-0287EA 265 EUR
552-0287 MANA814201 MANA814202 MANA814203 MANA814204 MANA814205 MANA814206
TLC education advanced kit, micro set F1
Lehrmittel Chemie-Lehrmittel Chromatographie-Lehrmittel
Advanced kit for science education.
  • All chemicals included for the separation of:
  • Amino acids test mixture and amino acids in urine sample
  • Heavy metal cations
Together with the materials from TLC micro-Set M an amino acid mixture (alanine, arginine, tryptophan, valine) as well as an heavy metal cation mixture (copper(II), manganese(II)) can be separated and visualised by spray reagents (ninhydrin and rubeanic acid).
Bitte beachten * These products contain harmful substances which must be specially labeled as hazardous.