Natriumhydroxid 2 N, BAKER ANALYZED® volumetrische Lösung, J.T.Baker®
Lieferant: Avantor
|
Gefahr
|
Synonyme:
Caustic soda
5633-02EA
0
EUR
5633-02
Natriumhydroxid 2 N, BAKER ANALYZED® volumetrische Lösung, J.T.Baker®
Sodium hydroxide
Standardisation at 25°C traceable to NIST Standard Reference Material.
Formel:
NaOH Molecular Weight: 40 g/mol Siedepunkt: 109 °C (1013 hPa) Dichte: ≤1,11 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003548 CAS-Nummer: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8,II |
Spezifikation Testergebnisse
For Laboratory,Research,or Manufacturing Use | 1.990 - 2.010 |
Protect from air to avoid absorption of carbon dioxide. | ≤5 ppm |
Normality | 1.990 - 2.010 |
Chloride (Cl) | ≤ 5 ppm |
ACS - Heavy Metals (as Pb) | ≤ 1 ppm |
Trace Impurities - Iron (Fe) | ≤ 0.5 ppm |
Standardization at 25°C traceable to NIST Standard Reference Material. | |
SRM NoÖ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...