ortho-Phosphorsäure ≥85%, Finyte® für Mikroelektronik, J.T.Baker®

Lieferant: Avantor

Synonyme: Phosphorsäure, Orthophosphorsäure

5854-05EA 516 EUR
5854-05
ortho-Phosphorsäure ≥85%, Finyte® für Mikroelektronik, J.T.Baker®
ortho-Phosphorsäure
Formel: H₃PO₄
Molecular Weight: 98 g/mol
MDL: MFCD00011340
CAS-Nummer: 7664-38-2
UN: 1805
ADR: 8,III

Bestellen

Spezifikation Testergebnisse

For Microelectronic Use 85.0 - 87.0 %
Store at temperatures above 22°C (72°F) = 10
Assay (H₃PO₄) (by acidimetry) 85.0 - 87.0 %
Color (APHA) ≤10
Volatile Acids (µeq/g) ≤0.16
Chloride (Cl) ≤0.7 ppm
Nitrate (NO₃) ≤1.5 ppm
Sulfate (SO₄) ≤8 ppm
Trace Impurities - Aluminum (Al) ≤400.0 ppb
Trace Impurities - Antimony (Sb) ≤6000.0 ppb
Trace Impurities - Arsenic (As) ≤50.0 ppb
Trace Impurities - Calcium (Ca) ≤450.0 ppb
Trace Impurities - Chromium (Cr) ≤200.0 ppb
Trace Impurities - Cobalt (Co) ≤50.0 ppb
Trace Impurities - Copper (Cu) ≤50 ppb
Trace Impurities - Gold (Au) ≤150.0 ppb
Trace Impurities - Iron (Fe) ≤700.0 ppb
Trace Impurities - Lead (Pb) ≤100 ppb
Trace Impurities - Lithium (Li) ≤100.0 ppb
Trace Impurities - Magnesium (Mg) ≤100.0 ppb
Trace Impurities - Manganese (Mn) ≤100.0 ppb
Trace Impurities - Nickel (Ni) ≤200.0 ppb
Trace Impurities - Potassium (K) ≤200.0 ppb
Trace Impurities - Sodium (Na) ≤500.0 ppb
Trace Impurities - Strontium (Sr) ≤100.0 ppb
Trace Impurities - Titanium (Ti) ≤300.0 ppb
Trace Impurities - Zinc (Zn) ≤300.0 ppb
Particle Count at point of fill - 0.5 µm and greater ≤50 par/ml
Particle Count at point of fill - 1.0 µm and greater ≤10 par/ml
For additional information, go to www.askavantor.com. Search keywords
freezing" and product name."

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR