Magnesiumsulfat, wasserfrei, Pulver, AR®, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials
Gefahr

Synonyme: MgSO4, Magnesium sulphate anhydrous

6070-26EA 0 EUR
6070-26
Magnesiumsulfat, wasserfrei, Pulver, AR®, Macron Fine Chemicals™
Magnesium sulphate
Formel: MgSO₄
Molecular Weight: 120,37 g/mol
Siedepunkt: 110 °C (1013 hPa)
Schmelzpunkt: 1127 °C
Dichte: 2,65 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011110
CAS-Nummer: 7487-88-9
EINECS: 231-298-2
ADR: 8,II
Merck Index: 12,05731
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bc2ae189b0cbc1bfc599951528466503d0d9008&vid=63867221020ebd2c8c227492e609a4764639ff79&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use
Ammonium (NH₄) ≤0.005 %
Arsenic (As) ≤0.001 %
Calcium (Ca) ≤0.04 %
Chloride (Cl) ≤0.001 %
Heavy Metals (as Pb) ≤0.001 %
Insoluble Matter ≤0.01 %
Trace Impurities - Iron (Fe) ≤0.001 %
Loss on Ignition ≤2.0 %
Thru U.S. No. 200 Sieve ≥80 %
Nitrate (NO₃) ≤0.005 %
pH of 5% Solution 5.0 - 9.2
Appearance (fine, white powder, free from lumps)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR