Analogue stopwatch, Lumen
Lieferant: HANHART
121.1515-00
609-1115EA 363 EUR
609-1115
Analogue stopwatch, Lumen
Timer Stoppuhren, analog
These watches, with luminous dials, are ideal for use in industry, laboratories and for athletes. Once the special dial has been 'charged' with light (takes approx. 1 hour), the face will glow in the dark for approx. 3 to 4 hours so it is clearly readable. This requires no additional energy or radiating materials. This is achieved by using an innovative plastic material containing light-storing pigments.
  • Robust ABS case with additional dust and water protection
  • Glass and glass fixing ring are moulded together and then firmly screwed to the case
  • Pin lever, 1 jewel