Analogue stopwatches, Amigo
Lieferant: HANHART
141.0434-00
609-1240EA 194 EUR
609-1240
Analogue stopwatches, Amigo
Timer Stoppuhren, analog
Analogue stopwatches with robust ABS casing.
  • Pin lever, one jewel
  • ¹/₁₀ second
  • 15 minutes capacity
  • Shock-protected pin lever movement
  • Robust ABS case, with cord for hanging round the neck