Jolting volumeter
Lieferant: GERBER FUNKE
7700
614-0255EA 1320 EUR
614-0255
Jolting volumeter
Dichtemessgeräte
This jolting volumeter is designed to determine the tamped volume or density of powdery and granular products.
Standards: ISO 8967 and IDF 134: Milk powder. USP 616 method I: Pharmaceutical powder. ISO 8460: Instant coffee.