Densimeter, DA-100M
Lieferant: Mettler - Toledo
51320001
614-1580EA 0 EUR
614-1580
Densimeter, DA-100M
Dichtemessgeräte
The DA-100M is a benchtop density meter of specific gravity or concentration.
  • Temperature control via built-in Peltier thermostat
  • Integrated air pump to dry the measuring cell
  • Average measuring time of 20 seconds
Suitable for measurements in aqueous solutions.