Chart for vaccine chart recorder, 4"
Lieferant: DICKSON
C204
620-1484EA 114 EUR
620-1484
Chart for vaccine chart recorder, 4"
Diagrammschreiber Diagrammscheiben für Schreiber
The vaccine chart recorder can be placed right inside the refrigerator or freezer along with the vaccines.
  • Easy-to-read special vaccine chart with circular bands highlight the acceptable vaccine ranges of 2 to 8 °C
  • LCD display shows current temperature for quick reference