Lebensmittelthermometer, TDC 110
Lieferant: EBRO
1340-5121
620-1688EA 30.3 EUR
620-1688
Lebensmittelthermometer, TDC 110
Thermometer Digitalthermometer
The low cost TDC 110 is an impact resistant thermometer suitable for fast temperature checks in the food industry, butchers and bakers. It provides measurements in air, liquids and soft materials.
  • LCD is 7 mm
  • Switchable scale between °C and °F
  • Automatic switch-off after 10 minutes of non use
Stainless steel fixed and pointed NTC probe (L×Ø: 120×4 mm).
Lieferumfang: Supplied with replaceable battery and needle guard in order to minimise the risk of injury.