Adapter, Dino-Lite FC-L-MA1
Lieferant: Dino-Lite / IDCP B.V.
FC-L-MA1
630-1817EA 56.5 EUR
630-1817
Adapter, Dino-Lite FC-L-MA1
Mikroskope
This mirror angle adapter fits onto the models with long working distance: AD4113TL/AD4116TL/AD4112PTL/AD4013MTL/AD4013MZTL/AD7013MTL and more.
Mirror angle: 45 °