Dow Corning® SYLGARD® 182 silicone elastomer
Lieferant: GREEFF RW SILICONES LUBRICAN
634153X
634153XEA 0 EUR
634153X
Dow Corning® SYLGARD® 182 silicone elastomer
Dow Corning® SYLGARD® 182 Silicone Elastomer
Dow Corning silicone 10:1 encapsulants for electronics are supplied in two parts as lot-matched base and curing agent that are mixed in a ratio of 10 parts base to one part curing agent, by weight.
  • Flexible and tranparent elastomer for easy board and part inspection
  • No exotherm during cure
  • No solvents or cure by-products
  • Deep section cure
  • Good dielectric properties
  • Easy to use and repair