Dow Corning® SYLGARD® 182 silicone elastomer
Lieferant: GREEFF RW SILICONES LUBRICAN
634153X
634153XEA
0
EUR
634153X
Dow Corning® SYLGARD® 182 silicone elastomer
Dow Corning® SYLGARD® 182 Silicone Elastomer
Dow Corning silicone 10:1 encapsulants for electronics are supplied in two parts as lot-matched base and curing agent that are mixed in a ratio of 10 parts base to one part curing agent, by weight.
- Flexible and tranparent elastomer for easy board and part inspection
- No exotherm during cure
- No solvents or cure by-products
- Deep section cure
- Good dielectric properties
- Easy to use and repair