Dow Corning® 732 Mehrzweck-Dichtstoff, schwarz
Lieferant: GREEFF RW SILICONES LUBRICAN
732B90
634272UEA 0 EUR
634272U
Dow Corning® 732 Mehrzweck-Dichtstoff, schwarz
Dow Corning 732 RTV Silicone Elastomer
General-purpose acetoxy RTV sealant. Meets the requirements of 21 CFR Section 177.2600: "Rubber articles intended for repeated use".
  • One-component adhesive/sealant
  • Cures at room temperature when exposed to moisture in the air
  • Acetoxy cure system
  • Non-sag, paste consistency, easy to apply
  • Cures to a tough, flexible rubber with good adhesion to many substrates
  • Excellent dielectric properties
Stable and flexible from −60 to +205 °C, with short peaks up to +230 °C
Complies with MIL-A-46106