Dowsil™ / Dow Corning 3145 Mil-A-46146 Haftdichtung, grau
Lieferant: GREEFF RW SILICONES LUBRICAN
634332P
634332PEA 0 EUR
634332P
Dowsil™ / Dow Corning 3145 Mil-A-46146 Haftdichtung, grau
Dow Corning® 3145 Mil-A-46146 Haftdichtung, grau
Non-flowing adhesive, high tensile strength and elongation, enhanced thermal stability, MIL-A-46146
  • One part
  • Polydimethylsiloxane adhesive
  • Room temperature cure, no ovens required
Lieferumfang: Supplied with nozzle