Digital Abbe refractometer
Lieferant: A.KRÜSS Optronic
AR2008
635-0086EA 7370 EUR
635-0086
Digital Abbe refractometer
Refraktometer Tischrefraktometer
The AR2008 is designed for applications in harsh environments.
  • Automatic temperature compensation (ATC)
  • Thermostat connection of prisms
  • Built-in light source for the measuring prism
The refractive index or Brix value and temperature is shown on a LCD display, with a serial interface allowing measured values to be transferred to a PC or printer.
Standards: ASTM D1218
Lieferumfang: Supplied with calibration plate, contact fluid and screw driver, as well as dust hood.