Pocket acidity meter, PAL-BX
Lieferant: ATAGO
7102
635-0729EA 581 EUR
635-0729
Pocket acidity meter, PAL-BX
Refraktometer Handrefraktometer
The PAL-BX pocket acidity meter is specifically for measuring citrus in grapes and wine.
  • Features with backlit screen
  • Accurate and easy measurements
  • Measure Brix and acidity within one unit
Standards: IP 65
Bestellinformation: Includes a digital scale, plastic pipettes, one beaker and one measuring spoon.