Abbe refractometer AR4
Lieferant: A.KRÜSS Optronic
AR4
635-2635EA 1570 EUR
635-2635
Abbe refractometer AR4
Refraktometer Tischrefraktometer
This Abbe refractometer is characterised by easy handling of small samples.
  • Readings via ocular
  • Adjustable scale, temperature controlled prisms with thermostat connections
  • For use with samples in the form of solids or pastes
Standards: ASTM D1218