n-Hexan 95%, ChromAR®, Macron Fine Chemicals™
Lieferant: Avantor Performance Materials
|
Gefahr
|
Synonyme:
Hexyl hydride, Hexanen, Hexane
6752-25EA
227.97
EUR
6752-25
n-Hexan 95%, ChromAR®, Macron Fine Chemicals™
n-Hexan
Formel:
H₃C(CH₂)₄CH₃ Molecular Weight: 86,18 g/mol |
MDL:
MFCD00009520 CAS-Nummer: 110-54-3 UN: 1208 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...