Tetramethylrhodamine methyl ester perchlorate (TMRM) ≥95% (durch HPLC) Mitochondrien-Färbemittel, geeignet für Fluoreszenz

Lieferant: Biotium
Achtung

Synonyme: N-[6-(Dimethylamino)-9-[2-(methoxycarbonyl)phenyl]-3H-xanthen-3-ylidene]-N-methylmethanaminium Perchlorate, Tetramethylrhodamine methyll ester Perchlorate, 3,6-Bis(dimethylamino)-9-(2-(methoxycarbonyl)phenyl)xanthylium perchlorate, Tetramethylrhodamine Methyl Ester Perchlorate

70017
70017.EA 342 EUR
70017.
Tetramethylrhodamine methyl ester perchlorate (TMRM) ≥95% (durch HPLC) Mitochondrien-Färbemittel, geeignet für Fluoreszenz
Tetramethylrhodamine methyl ester perchlorate (TMRM)

TMRM (Tetramethylrhodamine methyl ester, perchlorate) and TMRE are preferred dyes for quantitative measurements of membrane potentials using the Nernst equation.


  • Cellular localization: Mitochondria
  • For live or fixed cells: For live/intact cells
  • Assay type/options: Real-time imaging
  • Cell permeability: Membrane permeant
  • Apoptosis/viability marker: Mitochondrial potential
  • Potential dependence: Mitochondrial potential-dependent
  • Colors: Red
  • Excitation/Emission: 549/574 nm
  • Red solid soluble in DMSO, DMF or EtOH
  • Store at 4°C and protect from light


TMRM (Tetramethylrhodamine methyl ester, perchlorate) and TMRE (70016) are preferred dyes for quantitative measurements of membrane potentials using the Nernst equation. The dyes do not form aggregates in cell membranes and have minimal interaction with membrane proteins. Thus, the transmembrane distribution of the dyes is directly related to the membrane potential according to the Nernst equation. We also offer TMRM, 2 mM in DMSO.

Formel: C₂₅H₂₅N₂O₃·ClO₄
Molecular Weight: 500,93 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00274460
CAS-Nummer: 115532-50-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR