CSB-Bestimmungssysteme
Lieferant: GERHARDT
12-0060 12-0061 12-0073
705-0491EA 4125 EUR
705-0491 705-0492 705-0522
CSB-Bestimmungssysteme
Aufschlusssysteme Aufschluss-Systeme
The COD system is controlled by a digital electronic temperature controller to a precision of 1 K. The heating up phase of the the samples to ~148 °C is achieved in less than 10 minutes.
Beide Versionen sind mit einem Heizblock für 8 oder 20 Röhrchen verfügbar.
Manual version (M): The insert rack is handled manually in this version, which also includes 750 mm air condensers
Automatic version (A): This version includes a built-in motorised lift for the insert rack and a TZ controller for full automatic programming
The sytems contain the following components.
This version includes a built-in motorised lift for the insert rack and a TZ controller for full automatic programming.
The sytems contain the following components:
Bestellen