Portable multiparameter colorimeter, Orion™ AQUAfast™ AQ3700
Lieferant: Thermo Orion
AQ3700
705-0732EA 1660 EUR
705-0732
Portable multiparameter colorimeter, Orion™ AQUAfast™ AQ3700
Kolorimeter Mehrparameter-Kolorimeter
Measure common colorimetric water and wastewater parameters using the Orion™ AQUAfast™ AQ3700 colorimeter with 90 preprogrammed methods, automatic selection of 6 LED wavelengths and timer. Methods include total nitrogen and total phosphate nutrient testing or create up to 10 custom methods using 2-14 standards and blank.
  • Wide range of disinfectants in various formats to broaden our offering to a variety of industries
  • Large graphic display with step-by-step user instructions
  • Automatic countdown to measurement
  • Consistent results from shift to shift
  • Selectable languages of English, French, German, Italian, Spanish, Portuguese (Brazilian), and Polish with other languages uploadable via RJ 45 connection
Standards: FCC Part 15.