Kjeldahl distillation system, VAPODEST® 300
Lieferant: GERHARDT
12-0530
705-0858EA 0 EUR
705-0858
Kjeldahl distillation system, VAPODEST® 300
Destillationssysteme Kjeldahl-Destillationssysteme
VAPODEST® 300 is the ideal rapid water steam distllation instrument for Kjeldahl distillation. All glass components involved in the distillation process are back-lit for visual process control at any time.
  • Favourite method menu and easy menu navigation
  • 7" TFT full colour display
  • Programmable, automatic addition of water and NaOH
  • Programmable distillation time and reaction time
  • Programmable steam power 10 to 100% with steam soft start
  • Programmable suction of sample residue
VAPODEST® 300 complies with the data management requirements of accredited laboratories (ISO 17025, GLP).