Reflectometer, QUANTOFIX® Relax
Lieferant: MACHEREY-NAGEL
91346
705-1165EA 895 EUR
705-1165
Reflectometer, QUANTOFIX® Relax
Reflektometer
QUANTOFIX Relax is specially designed for test strip analyses.
  • High precision, quantitative results with no gradation and estimation of in-between values
  • Standardised reaction times
  • Easy data administration and quick testing procedure
Simplifies testing procedures and the decision processes by eliminating the subjectivity of visual readings.