Dimethylsulfoxid, BAKER ANALYZED®, Laborreagens, J.T.Baker®

Lieferant: Avantor

Synonyme: Methylsulfinylmethan, DMSO

7093.1000EA 0 EUR
7093.1000
Dimethylsulfoxid, BAKER ANALYZED®, Laborreagens, J.T.Baker®
Dimethylsulfoxid
Formel: (CH₃)₂SO
Molecular Weight: 78,14 g/mol
Siedepunkt: 189 °C (1013 hPa)
Schmelzpunkt: 18,5 °C
Dichte: 1,101 g/cm³ (20 °C)
Flammpunkt: 87 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002089
CAS-Nummer: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay min. 99.5 %
Color (APHA) max. 10
Residue after Evaporation max. 0.005 %
Water (H2O) max. 0.05 %
Ultraviolet Absorbance,(1.00-cm path vs water; curve smooth throughout stated range with no extraneous impurity peaks).
at 270 nm max. 1.0
at 275 nm max. 0.40
at 280 nm max. 0.30
at 300 nm max. 0.10
at 335 nm max. 0.03
at 400 nm max. 0.01

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR