Spreaders, L-shaped, plastic
Lieferant: Copan
174CS01 174CS05
710-0189EA 0 EUR
710-0189 710-0190
Spreaders, L-shaped, plastic
Schaber Zellschaber
These plastic, sterile L-shaped spreaders enable easy spreading of liquid samples across the surface of agar plates, without gouging or cutting the medium. Used for performing bacterial CFU plate counts on water, milk, urine and other liquid samples.
  • Foot with a completely smooth rounded surface, free of rough edges and imperfections
  • Pre-sterilised, eliminating the need for flaming or autoclaving