Spiral plater/diluter, easySpiral Dilute®
Lieferant: INTERSCIENCE
414000
710-2260EA 0 EUR
710-2260
Spiral plater/diluter, easySpiral Dilute®
Spiralplattengeräte
easySpiral Dilute® is a 2-in-1 equipment: automatic diluter and plater. It allows to do 5 x serial 1/10 dilutions and automatic plating on 1 single Petri dish, with a countable range of 30 to 10¹² CFU/ml.Controlled by microprocessor it is possible to use Petri dishes with diameter: 55, 90 and 150 mm.With sample mixing before dilution/plating and monthly cleaning program. Successive plating capacity with the same sample: 20 Petri dishes (50 μl).
  • One-touch panel for routine serial dilutions and plating
  • Make as many dilutions as you want
  • Fast: disinfection, 5 dilutions and 1 plating in 133 sec. 50% less consumables and time
  • According to ISO 7218 and Afnor V08-100
Preset dispensed volume: 50, 100 or 200 µl
Lieferumfang: Delivered with 1000 sterile beakers, 1 blue dye for testing, 1 liquid detergent, 3 sets of GL45 bottle connectors, Spiral® counting grid: 90 and 150 mm, circle mode counting grid: 90 and 150 mm, 55 mm Petri dish ring, 90 mm Petri dish ring, 150 mm Petri dish ring, software on a CD-ROM, USB cable, power cable, user manual.