Natriumhydroxid

Lieferant: WTW
Gefahr

Synonyme: Caustic soda

209140
715-3224EA 68 EUR
715-3224
Natriumhydroxid
Sodium hydroxide

Sodium hydroxide pellets is used as the carbon dioxide absorbent for BOD measurements


  • Product category: PC21 laboratory chemicals
  • Process category: PROC15 use as laboratory reagent

Formel: NaOH
Molecular Weight: 40 g/mol
Siedepunkt: 1390 °C (1013 hPa)
Schmelzpunkt: 319 °C
Dichte: 2,13 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003548
CAS-Nummer: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
REACH: 01-2119457892-27
Merck Index: 14,08627

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR