Sodium ionophore II, Selectophore™, Funktion getestet Ionophor, Supelco®

Lieferant: Merck

Synonyme: 2,2'-(1,2-Phenylenebis(oxy))bis(N-benzyl-N-phenylacetamide), N,N′-Dibenzyl-N,N′-diphenyl-1,2-phenylenedioxydiacetamide, ETH 157

71733-25MG
71733-25MGEA 245 EUR
71733-25MG
Sodium ionophore II, Selectophore™, Funktion getestet Ionophor, Supelco®
Sodium ionophore II

Sodium ionophore II is commonly used sodium selective neutral carrier having 2.5 times more sensitivity towards sodium ions than to potassium ions. It can be used in ion-selective electrode preparation


Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.

Formel: C₃₆H₃₂N₂O₄
Molecular Weight: 556,65 g/mol
MDL: MFCD00042979
CAS-Nummer: 61595-77-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9aa75d16cf0395337a81330be1478c358a301f9e&vid=62e1199a21c6435e2c73b0c91736ddc8141fb4d7&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR