Reader, Micro-Haematocrit
Lieferant: HAWKSLEY AND SONS
01502-00
720-0100EA 263 EUR
720-0100
Reader, Micro-Haematocrit
Klinisches Diagnosezubehör Klinische Analysegeräte
For determination of packed cell volume, using 75 mm capillary tubes with total fluid column heights between 40 and 70 mm.