Capillary blood sampling system, Capiject
Lieferant: TERUMO
CT-M T-MGA CT-MLH CT-MPS CT-MQK CT-MG CT-MLHG
720-2107EA 167 EUR
720-2107 720-2109 720-2110 720-2112 720-2113 720-2108 720-2111
Capillary blood sampling system, Capiject
Pipetten Mikrokapillaren
Extra wide collection lip for enhanced specimen flow and reduced contamination.
  • Accurate and easy sampling
  • The sleek, tapered siliconecoated interior additionally reduces sticking
  • The ridged cap facilitates opening and closure
  • Colour coded fill-lines
  • Superior olefin-based gel for clean separations
  • Batch number and shelf-life on each tube