Salzsäure 10%, BAKER, Laborreagens

Lieferant: Avantor
Achtung

Synonyme: Hydrochloric acid solution, Muriatic acid

7219.9060EA 588.92 EUR
7219.9060
Salzsäure 10%, BAKER, Laborreagens
Salzsäure
Formel: HCl
Molecular Weight: 36,46 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011324
CAS-Nummer: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

Assay 9.5 - 10.5 %
Appearance passes test
Free Chlorine max. 1 ppm
Heavy Metals (as Pb) max. 2 ppm
Identification passes test
Residue after Evaporation max. 0.01 %
Sulfates (as SO₄) max. 5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR