Transfer buffer, Tris-glycine,10X, AccuGENE™
Lieferant: LONZA
50879
733-1629EA
197
EUR
733-1629
Transfer buffer, Tris-glycine,10X, AccuGENE™
Puffer
Elektrophorese-Puffer
Transferpuffer
AccuGENE™ electrophoresis buffers are ready-to-use solutions ideal for a wide range of molecular biology applications.
- Ready to use solutions
- Reliable manufactured according to strict quality control standards to ensure lot to lot consistency
- Guaranteed DNase-, RNase- and protease-free
Each reagent is prepared with 18 megOhm water, filtered using a 0,2 micron filter, and filled into sterile bottles.