Deckel für 96-Well-PCR-Platte, Corning® Thermowell®
Lieferant: Corning
6520
734-1754EA 136.69 EUR
734-1754
Deckel für 96-Well-PCR-Platte, Corning® Thermowell®
Deckel Deckel für Mikrotiterplatten
Polycarbonate lids are thermally stable up to 135 °C and can be used on Corning® Thermowell™ 96 well polycarbonate plates (fits Cat. No. 6511) with oil overlays.
Lids are intended to be covers only and do not prevent evaporation.